siRNA Details
| Gene Symbol: KIFC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145754.3 | NP_665697.1 | 11, 12 | 1312 | ||
| XM_005272357.3 | XP_005272414.1 | 11, 12 | 1402 | |||
| XM_011517362.2 | XP_011515664.1 | 11, 12 | 1403 | |||
| GenBank mRNA | AK055045.1 | - | 8, 9 | 690 | ||
| BC017311.2 | - | 12, 13 | 1294 | 5090622 | ||

KIFC2
Genomic Map