siRNA Details
| Gene Symbol: TTC21A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105513.2 | NP_001098983.2 | 18 | 2576 | ||
| NM_145755.2 | NP_665698.2 | 19 | 2720 | |||
| XM_005264921.4 | XP_005264978.1 | 19 | 2649 | |||
| XM_005264922.4 | XP_005264979.1 | 19 | 2649 | |||
| XM_005264923.3 | XP_005264980.1 | 19 | 2649 | |||
| XM_005264924.4 | XP_005264981.1 | 18 | 2538 | |||
| XM_005264925.4 | XP_005264982.1 | 18 | 2526 | |||
| XM_005264927.4 | XP_005264984.1 | 15 | 2103 | |||
| XM_006713011.3 | XP_006713074.1 | 19 | 2649 | |||
| XM_011533447.2 | XP_011531749.1 | 18 | 2550 | |||
| XM_011533449.1 | XP_011531751.1 | 17 | 2399 | |||
| GenBank mRNA | AJ487015.1 | - | 19 | 2682 | ||
| AK093313.1 | - | 9 | 1260 | |||
| AK302408.1 | - | 18 | 2559 | |||
| AK302465.1 | - | 18 | 2430 | |||
| BC129948.1 | - | 19 | 2631 | 40126472 | ||

TTC21A
Genomic Map