siRNA Details
| Gene Symbol: ZNF396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322286.1 | NP_001309215.1 | 4 | 3577 | ||
| NM_001322290.1 | NP_001309219.1 | 5 | 3814 | |||
| NM_145756.2 | NP_665699.1 | 5 | 2707 | |||
| GenBank mRNA | AK055775.1 | - | 5 | 2711 | ||

ZNF396
Genomic Map