siRNA Details
| Gene Symbol: ABTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145804.2 | NP_665803.2 | 17 | 3915 | ||
| GenBank mRNA | AK056863.1 | - | 5 | 971 | ||
| AK091560.1 | - | 14 | 2133 | |||
| AK095632.1 | - | 17 | 3162 | |||
| AL050374.1 | - | 2 | 687 | |||
| BX640841.1 | - | 9 | 1616 | |||

ABTB2
Genomic Map