siRNA Details
| Gene Symbol: ANKRD18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006716749.3 | XP_006716812.1 | 17, 18 | 3361 | ||
| XM_011517836.2 | XP_011516138.1 | 17, 18 | 3362 | |||
| XM_011517837.2 | XP_011516139.1 | 17, 18 | 3362 | |||
| GenBank mRNA | AK093059.1 | - | 3, 4 | 420 | ||
| AY550933.1 | - | 3, 4 | 369 | |||
| BC131497.1 | - | 3, 4 | 416 | |||

ANKRD18A
Genomic Map