siRNA Details
| Gene Symbol: BOD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148894.2 | NP_683692.2 | 3 | 498 | ||
| XM_005248150.3 | XP_005248207.1 | 3 | 515 | |||
| XM_005248151.3 | XP_005248208.1 | 3 | 515 | |||
| XM_006713958.3 | XP_006714021.1 | 3 | 515 | |||
| XM_011513827.2 | XP_011512129.1 | 3 | 515 | |||
| XM_011513829.2 | XP_011512131.1 | 3 | 515 | |||
| XM_017008009.1 | XP_016863498.1 | 3 | 515 | |||
| XM_017008010.1 | XP_016863499.1 | 3 | 515 | |||
| GenBank mRNA | AF528529.1 | - | 3 | 498 | ||
| BC065546.1 | - | 3 | 447 | |||
| BC087835.1 | - | 3 | 504 | |||

BOD1L1
Genomic Map