siRNA Details
| Gene Symbol: HUS1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148959.3 | NP_683762.2 | 1 | 616 | ||
| GenBank mRNA | AF508547.1 | - | 1 | 596 | ||
| BC101697.1 | - | 1 | 616 | 8069203 | ||
| BC101699.1 | - | 1 | 616 | 8069205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HUS1B| Gene Symbol: HUS1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148959.3 | NP_683762.2 | 1 | 616 | ||
| GenBank mRNA | AF508547.1 | - | 1 | 596 | ||
| BC101697.1 | - | 1 | 616 | 8069203 | ||
| BC101699.1 | - | 1 | 616 | 8069205 | ||