siRNA Details
Gene Symbol: PSMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143937.1 | NP_001137409.1 | 2, 3 | 202 | ||
NM_002786.3 | NP_002777.1 | 2, 3 | 202 | |||
NM_148976.2 | NP_683877.1 | 3, 4 | 419 | |||
GenBank mRNA | AK290765.1 | - | 2, 3 | 143 | ||
AK303565.1 | - | 2, 3 | 139 | |||
BC002577.2 | - | 2, 3 | 121 | 3140784 | ||
BC005932.1 | - | 2, 3 | 137 | 4078894 | ||
BC008472.1 | - | 2, 3 | 102 | 4282076 | ||
BC009576.1 | - | 2, 3 | 110 | 4105838 | ||
BC015105.1 | - | 2, 3 | 136 | 3950568 | ||
BC015356.1 | - | 2, 3 | 105 | 3452219 | ||
BC022372.1 | - | 2, 3 | 89 | 4770620 | ||
BT006647.1 | - | 1, 2 | 56 | |||
D00759.1 | - | 2, 3 | 97 | |||
DA549688.1 | - | 3, 4 | 509 | |||
FJ224300.1 | - | 3, 4 | 401 | |||
M64992.1 | - | 3, 4 | 401 | |||
X61969.1 | - | 2, 3 | 161 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
