siRNA Details
| Gene Symbol: IPMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152230.4 | NP_689416.1 | 1, 2 | 511 | ||
| XM_011539565.2 | XP_011537867.1 | 1, 2 | 543 | |||
| GenBank mRNA | AF432853.1 | - | 1, 2 | 188 | ||
| BC065709.1 | - | 1, 2 | 515 | 6654071 | ||

IPMK
Genomic Map