siRNA Details
| Gene Symbol: SNX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021249.3 | NP_067072.3 | 2 | 256 | ||
| NM_152233.2 | NP_689419.2 | 3 | 151 | |||
| XM_006720224.1 | XP_006720287.1 | 3 | 149 | |||
| GenBank mRNA | AF121856.1 | - | 3 | 142 | ||
| AK292250.1 | - | 3 | 134 | |||
| AK296213.1 | - | 3 | 145 | |||
| AK308464.1 | - | 3 | 145 | |||
| BC001798.1 | - | 2 | 256 | 3354467 | ||
| BG722853.1 | - | 3 | 177 | |||
| U83194.1 | - | 3 | 119 | |||

SNX6
Genomic Map