siRNA Details
| Gene Symbol: FAAP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152266.4 | NP_689479.1 | 2, 3 | 247 | ||
| GenBank mRNA | AK128668.1 | - | 2, 3 | 214 | ||
| BC003535.1 | - | 2, 3 | 167 | 3609326 | ||
| BC010170.2 | - | 2, 3 | 175 | 3659284 | ||
| BC020247.1 | - | 2, 3 | 195 | 4620233 | ||
| DA006539.1 | - | 2, 3 | 222 | |||
| KU178830.1 | - | 1, 2 | 104 | |||

FAAP24
Genomic Map