siRNA Details
| Gene Symbol: TBL1X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139466.1 | NP_001132938.1 | 18 | 2249 | ||
| NM_001139467.1 | NP_001132939.1 | 17 | 2104 | |||
| NM_001139468.1 | NP_001132940.1 | 18 | 2239 | |||
| NM_005647.3 | NP_005638.1 | 18 | 2368 | |||
| XM_011545571.2 | XP_011543873.1 | 17 | 2090 | |||
| GenBank mRNA | AB209608.1 | - | 13 | 2322 | ||
| BC032708.1 | - | 18 | 2215 | 5496758 | ||
| BC052304.1 | - | 18 | 2368 | 6378852 | ||
| Y12781.1 | - | 18 | 2527 | |||

TBL1X
Genomic Map