siRNA Details
| Gene Symbol: DMBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147192.2 | NP_671725.1 | 4 | 1249 | ||
| NM_172225.1 | NP_757379.1 | 4 | 1234 | |||
| XM_011540668.2 | XP_011538970.1 | 5 | 1480 | |||
| XM_017000289.1 | XP_016855778.1 | 5 | 1272 | |||
| GenBank mRNA | AB037699.1 | - | 4 | 1234 | ||

DMBX1
Genomic Map