siRNA Details
| Gene Symbol: SMC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291501.1 | NP_001278430.1 | 3, 4 | 471 | ||
| NM_148674.4 | NP_683515.4 | 3, 4 | 471 | |||
| XM_011530144.2 | XP_011528446.1 | 3, 4 | 332 | |||
| XM_011530145.2 | XP_011528447.1 | 3, 4 | 523 | |||
| GenBank mRNA | AJ504806.2 | - | 3, 4 | 471 | ||
| AL704843.1 | - | 3, 4 | 450 | |||
| BC126208.1 | - | 3, 4 | 451 | 8991924 | ||

SMC1B
Genomic Map