siRNA Details
| Gene Symbol: LONRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152271.3 | NP_689484.3 | 10 | 1978 | ||
| XM_005273685.3 | XP_005273742.1 | 10 | 1947 | |||
| XM_011544694.2 | XP_011542996.1 | 10 | 1730 | |||
| GenBank mRNA | AK074329.1 | - | 5 | 530 | ||
| AK297266.1 | - | 9 | 1115 | |||
| AK300538.1 | - | 6 | 724 | |||
| BC004538.2 | - | 2 | 114 | |||
| BC106042.1 | - | 10 | 1244 | |||
| BC117381.1 | - | 7 | 909 | 40125932 | ||
| BC117385.1 | - | 7 | 909 | 40125936 | ||

LONRF1
Genomic Map