siRNA Details
| Gene Symbol: FAM58A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152274.4 | NP_689487.2 | 4, 5 | 769 | ||
| XM_005277920.4 | XP_005277977.1 | 4, 5 | 727 | |||
| XM_011531214.2 | XP_011529516.1 | 4, 5 | 1293 | |||
| XM_011531215.2 | XP_011529517.1 | 4, 5 | 1159 | |||
| GenBank mRNA | BC001909.1 | - | 4, 5 | 709 | 3537447 | |
| BC007232.1 | - | 4, 5 | 709 | 3347302 | ||
| BC032121.2 | - | 4, 5 | 717 | 4575331 | ||
| BC071851.1 | - | 4, 5 | 570 | 5934127 | ||

FAM58A
Genomic Map