siRNA Details
| Gene Symbol: CLRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152311.3 | NP_689524.1 | 3 | 801 | ||
| XM_011539274.2 | XP_011537576.1 | 2 | 620 | |||
| GenBank mRNA | BC029478.1 | - | 3 | 801 | 4733535 | |
| BM830163.1 | - | 3 | 554 | |||
| BX640823.1 | - | 2 | 1081 | |||

CLRN3
Genomic Map