siRNA Details
| Gene Symbol: FAM110B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147189.2 | NP_671722.1 | 5 | 2645 | ||
| XM_005251324.2 | XP_005251381.1 | 4 | 2639 | |||
| XM_005251325.3 | XP_005251382.1 | 3 | 12802 | |||
| XM_005251326.2 | XP_005251383.1 | 3 | 12827 | |||
| XM_017013948.1 | XP_016869437.1 | 4 | 5214 | |||
| GenBank mRNA | BC024294.1 | - | 2 | 2157 | 5440447 | |

FAM110B
Genomic Map