siRNA Details
Gene Symbol: WFDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147197.2 | NP_671730.1 | 5 | 521 | ||
GenBank mRNA | AY047609.1 | - | 5 | 521 | ||
BC062670.1 | - | 5 | 552 | 6101449 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WFDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147197.2 | NP_671730.1 | 5 | 521 | ||
GenBank mRNA | AY047609.1 | - | 5 | 521 | ||
BC062670.1 | - | 5 | 552 | 6101449 |