siRNA Details
| Gene Symbol: IPMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152230.4 | NP_689416.1 | 4, 5 | 876 | ||
| XM_011539565.2 | XP_011537867.1 | 4, 5 | 917 | |||
| XM_017016013.1 | XP_016871502.1 | 4, 5 | 543 | |||
| GenBank mRNA | AF432853.1 | - | 4, 5 | 553 | ||
| AK096839.1 | - | 1, 2 | 183 | |||
| BC065709.1 | - | 4, 5 | 880 | 6654071 | ||

IPMK
Genomic Map