siRNA Details
| Gene Symbol: ZNF585B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152279.3 | NP_689492.3 | 5 | 1091 | ||
| GenBank mRNA | AK091586.1 | - | 2 | 265 | ||
| AK094989.1 | - | 1 | 1093 | |||
| AY147015.1 | - | 5 | 1093 | |||
| BC080550.1 | - | 6 | 856 | |||
| BC093625.1 | - | 4 | 933 | 7939470 | ||

ZNF585B
Genomic Map