siRNA Details
| Gene Symbol: C1orf158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152290.3 | NP_689503.3 | 4 | 955 | ||
| XM_011542434.1 | XP_011540736.1 | 3 | 826 | |||
| GenBank mRNA | AK026705.1 | - | 4 | 923 | ||
| BC029894.1 | - | 4 | 926 | 5171302 | ||
| BX647383.1 | - | 1024 | ||||

C1orf158
Genomic Map