siRNA Details
| Gene Symbol: ZNF554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102651.1 | NP_001096121.1 | 5 | 2096 | ||
| XM_006722635.3 | XP_006722698.1 | 4 | 1783 | |||
| XM_011527664.2 | XP_011525966.1 | 3 | 3068 | |||
| XM_017026235.1 | XP_016881724.1 | 3 | 3413 | |||
| XM_017026236.1 | XP_016881725.1 | 3 | 1781 | |||
| GenBank mRNA | AK092136.1 | - | 6 | 2148 | ||
| BC000113.1 | - | 5 | 2007 | |||

ZNF554
Genomic Map