siRNA Details
| Gene Symbol: KRT222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152349.2 | NP_689562.1 | 1, 2 | 162 | ||
| GenBank mRNA | BC032815.1 | - | 1, 2 | 121 | 4346009 | |
| DB465511.1 | - | 1, 2 | 162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT222| Gene Symbol: KRT222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152349.2 | NP_689562.1 | 1, 2 | 162 | ||
| GenBank mRNA | BC032815.1 | - | 1, 2 | 121 | 4346009 | |
| DB465511.1 | - | 1, 2 | 162 | |||