siRNA Details
| Gene Symbol: FAM210A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098801.1 | NP_001092271.1 | 3 | 539 | ||
| NM_152352.3 | NP_689565.2 | 2 | 399 | |||
| GenBank mRNA | AK055618.1 | - | 3 | 495 | ||
| DA560663.1 | - | 3 | 469 | |||
| DA895646.1 | - | 2 | 369 | |||
| DA984075.1 | - | 2 | 396 | |||

FAM210A
Genomic Map