siRNA Details
| Gene Symbol: SGPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152386.3 | NP_689599.2 | 1, 2 | 323 | ||
| GenBank mRNA | AF542512.1 | - | 1, 2 | 227 | ||
| AK314322.1 | - | 1, 2 | 323 | |||
| BC106904.1 | - | 1, 2 | 218 | |||
| BC106905.1 | - | 1, 2 | 220 | |||
| BC134342.1 | - | 1, 2 | 227 | 40031702 | ||

SGPP2
Genomic Map