siRNA Details
| Gene Symbol: TMEM68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286657.1 | NP_001273586.1 | 2 | 227 | ||
| NM_001286660.1 | NP_001273589.1 | 3 | 366 | |||
| NM_152417.2 | NP_689630.1 | 3 | 366 | |||
| XM_005251150.1 | XP_005251207.1 | 3 | 355 | |||
| XM_011517454.1 | XP_011515756.1 | 3 | 379 | |||
| XM_011517455.2 | XP_011515757.1 | 3 | 355 | |||
| GenBank mRNA | AK056932.1 | - | 3 | 285 | ||
| AK308247.1 | - | 3 | 269 | |||
| BC020835.1 | - | 3 | 325 | 4806471 | ||
| BC071636.1 | - | 3 | 326 | 5193338 | ||

TMEM68
Genomic Map