siRNA Details
| Gene Symbol: HGSNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152419.2 | NP_689632.2 | 14, 15 | 1520 | ||
| XM_005273409.1 | XP_005273466.1 | 15, 16 | 1631 | |||
| XM_005273411.1 | XP_005273468.1 | 13, 14 | 1439 | |||
| XM_017013088.1 | XP_016868577.1 | 12, 13 | 1328 | |||
| XM_017013089.1 | XP_016868578.1 | 13, 14 | 1393 | |||
| GenBank mRNA | AK057293.1 | - | 5, 6 | 910 | ||
| AK074839.1 | - | 1, 2 | 90 | |||
| AK304441.1 | - | 14, 15 | 1472 | |||
| AK308922.1 | - | 1 | 788 | |||
| CR749838.1 | - | 10, 11 | 944 | |||

HGSNAT
Genomic Map