siRNA Details
| Gene Symbol: ZNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006965.3 | NP_008896.2 | 4 | 1522 | ||
| XM_005258341.3 | XP_005258398.1 | 4 | 1509 | |||
| GenBank mRNA | AF038964.1 | - | 4 | 1466 | ||
| AK291246.1 | - | 4 | 1501 | |||
| U68536.1 | - | 4 | 1496 | |||

ZNF24
Genomic Map