siRNA Details
| Gene Symbol: MTERF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301134.1 | NP_001288063.1 | 2 | 1557 | ||
| NM_001301135.1 | NP_001288064.1 | 4 | 1717 | |||
| NM_006980.4 | NP_008911.1 | 3 | 1616 | |||
| XM_005250593.3 | XP_005250650.1 | 3 | 1607 | |||
| XM_006716126.3 | XP_006716189.1 | 3 | 1646 | |||
| XM_017012620.1 | XP_016868109.1 | 4 | 1624 | |||
| GenBank mRNA | AK222730.1 | - | 3 | 1616 | ||
| AL832861.1 | - | 2 | 1527 | |||
| BC000965.3 | - | 2 | 1552 | |||
| Y09615.1 | - | 3 | 1599 | |||

MTERF1
Genomic Map