siRNA Details
| Gene Symbol: LOC652276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015441.1 | 3, 4 | 1048 | |||
| GenBank mRNA | AK096394.1 | - | 3, 4 | 1082 | ||
| AK291606.1 | - | 3, 4 | 1032 | |||
| BC062576.1 | - | 3, 4 | 1012 | |||
| BC080650.1 | - | 3, 4 | 978 | |||

LOC652276
Genomic Map