siRNA Details
| Gene Symbol: PAQR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284511.1 | NP_001271440.1 | 3 | 1641 | ||
| NM_001284512.1 | NP_001271441.1 | 2 | 1536 | |||
| NM_001284513.1 | NP_001271442.1 | 3 | 1264 | |||
| NM_001324118.1 | NP_001311047.1 | 4 | 1557 | |||
| NM_152341.4 | NP_689554.2 | 3 | 1758 | |||
| GenBank mRNA | AK054564.1 | - | 3 | 1342 | ||
| AK092173.1 | - | 2 | 1223 | |||
| AL832527.1 | - | 3 | 1371 | |||
| AY424282.1 | - | 3 | 1328 | |||
| BC033703.1 | - | 3 | 1416 | 4154019 | ||

PAQR4
Genomic Map