siRNA Details
| Gene Symbol: ZNF491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152356.3 | NP_689569.2 | 3 | 1099 | ||
| XM_005259730.3 | XP_005259787.1 | 2 | 1040 | |||
| GenBank mRNA | AK092110.1 | - | 1 | 583 | ||
| AK096593.1 | - | 3 | 1091 | |||
| AK297479.1 | - | 4 | 1045 | |||
| BC101590.1 | - | 2 | 912 | 8069096 | ||
| BC101592.1 | - | 2 | 912 | 8069098 | ||

ZNF491
Genomic Map