siRNA Details
| Gene Symbol: MYOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152372.3 | NP_689585.3 | 29 | 3612 | ||
| GenBank mRNA | AK093280.1 | - | 2 | 498 | ||
| AK128233.1 | - | 21 | 2672 | |||
| BC014671.1 | - | 3 | 132 | |||
| BC067101.1 | - | 29 | 3622 | 5928128 | ||
| BX537911.1 | - | 28 | 3336 | |||
| BX647453.1 | - | 29 | 3573 | |||
| BX647455.1 | - | 29 | 3612 | |||
| BX647644.1 | - | 29 | 3573 | |||

MYOM3
Genomic Map