siRNA Details
| Gene Symbol: TMEM237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044385.2 | NP_001037850.1 | 5 | 368 | ||
| NM_152388.3 | NP_689601.2 | 5 | 533 | |||
| GenBank mRNA | AB053301.1 | - | 5 | 535 | ||
| AK090601.1 | - | 5 | 226 | |||
| AK303954.1 | - | 5 | 340 | |||
| BC029611.1 | - | 5 | 281 | 5304046 | ||
| BP226834.1 | - | 5 | 368 | |||
| BQ219059.1 | - | 5 | 340 | |||

TMEM237
Genomic Map