siRNA Details
| Gene Symbol: EMILIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007046.3 | NP_008977.1 | 8 | 3915 | ||
| GenBank mRNA | AF088916.1 | - | 8 | 3416 | ||
| AK075517.1 | - | 5 | 2097 | |||
| AK307539.1 | - | 6 | 2031 | |||
| AL050138.1 | - | 8 | 3346 | |||
| BC007530.1 | - | 9 | 2373 | |||
| BC009947.2 | - | 5 | 2110 | |||
| BC142638.1 | - | 8 | 2643 | |||
| BC142688.1 | - | 8 | 3797 | 40148654 | ||

EMILIN1
Genomic Map