siRNA Details
| Gene Symbol: VEZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007146.2 | NP_009077.2 | 4 | 1009 | ||
| XM_005257643.1 | XP_005257700.1 | 4 | 1006 | |||
| XM_005257644.4 | XP_005257701.1 | 5 | 1162 | |||
| XM_017025017.1 | XP_016880506.1 | 5 | 1213 | |||
| XM_017025018.1 | XP_016880507.1 | 5 | 1065 | |||
| GenBank mRNA | BX648862.1 | - | 3 | 326 | ||
| D28118.1 | - | 4 | 907 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_016686, AF104410.

VEZF1
Genomic Map