siRNA Details
| Gene Symbol: SUPT16H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007192.3 | NP_009123.1 | 5 | 944 | ||
| XM_011536381.2 | XP_011534683.1 | 6 | 946 | |||
| GenBank mRNA | AK001719.1 | - | 5 | 680 | ||
| AK024072.1 | - | 5 | 946 | |||
| BC000565.2 | - | 5 | 681 | |||
| BC014046.2 | - | 5 | 681 | |||
| BC064561.1 | - | 5 | 669 | |||
| BC073849.1 | - | 5 | 678 | |||
| BC111402.1 | - | 5 | 693 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_033618, AK158116, AK146529, AK148506, AI930189, and others.

SUPT16H
Genomic Map