siRNA Details
| Gene Symbol: ZG16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152338.3 | NP_689551.2 | 4 | 619 | ||
| XM_011545921.2 | XP_011544223.1 | 4 | 698 | |||
| GenBank mRNA | AK125559.1 | - | 4 | 577 | ||
| BC029149.1 | - | 3 | 551 | 5187790 | ||
| BC150656.1 | - | 4 | 590 | 9057027 | ||

ZG16
Genomic Map