siRNA Details
| Gene Symbol: ZNF491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152356.3 | NP_689569.2 | 3 | 1723 | ||
| XM_005259730.3 | XP_005259787.1 | 2 | 1664 | |||
| GenBank mRNA | AK092110.1 | - | 1 | 1207 | ||
| AK096593.1 | - | 3 | 1715 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF491| Gene Symbol: ZNF491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152356.3 | NP_689569.2 | 3 | 1723 | ||
| XM_005259730.3 | XP_005259787.1 | 2 | 1664 | |||
| GenBank mRNA | AK092110.1 | - | 1 | 1207 | ||
| AK096593.1 | - | 3 | 1715 | |||