siRNA Details
| Gene Symbol: SGPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320833.1 | NP_001307762.1 | 6 | 1312 | ||
| NM_001320834.1 | NP_001307763.1 | 5 | 881 | |||
| NM_152386.3 | NP_689599.2 | 5 | 1169 | |||
| XM_017003355.1 | XP_016858844.1 | 5 | 1071 | |||
| GenBank mRNA | AF542512.1 | - | 5 | 1073 | ||
| AK096323.1 | - | 5 | 883 | |||
| AK314322.1 | - | 5 | 1169 | |||
| BC106904.1 | - | 5 | 1064 | |||
| BC106905.1 | - | 5 | 1066 | |||
| BC134342.1 | - | 5 | 1073 | 40031702 | ||

SGPP2
Genomic Map