siRNA Details
| Gene Symbol: JMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152405.4 | NP_689618.4 | 11 | 4024 | ||
| XM_005248430.2 | XP_005248487.1 | 11 | 3901 | |||
| XM_011543155.2 | XP_011541457.1 | 10 | 3928 | |||
| GenBank mRNA | AK095189.1 | - | 10 | 2425 | ||
| AL708168.1 | - | 2 | 591 | |||
| BX648731.1 | - | 3 | 1222 | |||

JMY
Genomic Map