siRNA Details
| Gene Symbol: TMEM68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286657.1 | NP_001273586.1 | 7 | 1156 | ||
| NM_001286661.1 | NP_001273590.1 | 5 | 716 | |||
| NM_152417.2 | NP_689630.1 | 6 | 1094 | |||
| XM_005251150.1 | XP_005251207.1 | 8 | 1284 | |||
| XM_011517454.1 | XP_011515756.1 | 8 | 1308 | |||
| XM_017013040.1 | XP_016868529.1 | 7 | 868 | |||
| GenBank mRNA | AK056932.1 | - | 6 | 1013 | ||
| AL832935.1 | - | 6 | 765 | |||
| BC043417.1 | - | 4 | 3256 | |||

TMEM68
Genomic Map