siRNA Details
| Gene Symbol: CARNMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320497.1 | NP_001307426.1 | 8 | 1330 | ||
| NM_152420.2 | NP_689633.1 | 8 | 1650 | |||
| NR_135282.1 | 7 | 1509 | ||||
| XM_005251701.3 | XP_005251758.2 | 8 | 1325 | |||
| XM_017014269.1 | XP_016869758.1 | 8 | 1637 | |||
| XM_017014270.1 | XP_016869759.1 | 9 | 1583 | |||
| XM_017014271.1 | XP_016869760.1 | 9 | 2075 | |||
| GenBank mRNA | BC034033.1 | - | 8 | 1614 | 4278547 | |
| BX538061.1 | - | 7 | 1167 | |||
| BX647386.1 | - | 8 | 1419 | |||

CARNMT1
Genomic Map