siRNA Details
| Gene Symbol: GTF3C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206774.1 | NP_001193703.1 | 2 | 337 | ||
| NM_012086.4 | NP_036218.1 | 2 | 337 | |||
| XM_005246965.4 | XP_005247022.1 | 2 | 252 | |||
| GenBank mRNA | AF133123.1 | - | 2 | 273 | ||
| AF465407.1 | - | 2 | 228 | |||
| AK025560.1 | - | 2 | 251 | |||
| AK314718.1 | - | 2 | 278 | |||
| BC043347.1 | - | 2 | 192 | 5498609 | ||
| DB078021.1 | - | 2 | 337 | |||

GTF3C3
Genomic Map