siRNA Details
| Gene Symbol: KBTBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152433.3 | NP_689646.2 | 3 | 1356 | ||
| NM_198439.2 | NP_940841.1 | 4 | 1557 | |||
| XM_006718767.3 | XP_006718830.1 | 4 | 1587 | |||
| XM_011542618.2 | XP_011540920.1 | 3 | 1201 | |||
| XM_011542619.2 | XP_011540921.1 | 3 | 1339 | |||
| GenBank mRNA | AK092993.1 | - | 3 | 959 | ||
| AK289915.1 | - | 4 | 1167 | |||
| BC117466.1 | - | 3 | 1055 | 40126017 | ||
| BX640672.1 | - | 3 | 1077 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026962, NM_001164574, AK034334, AK083497, BC027392, and others.

KBTBD3
Genomic Map