siRNA Details
| Gene Symbol: TMCO5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152453.3 | NP_689666.2 | 3 | 320 | ||
| XM_005254168.3 | XP_005254225.1 | 3 | 301 | |||
| XM_011521262.2 | XP_011519564.1 | 3 | 301 | |||
| XM_011521263.1 | XP_011519565.1 | 3 | 301 | |||
| XM_011521264.1 | XP_011519566.1 | 3 | 301 | |||
| GenBank mRNA | AK093126.1 | - | 3 | 301 | ||
| AK310517.1 | - | 3 | 306 | |||
| BC029221.2 | - | 2 | 233 | 5167874 | ||
| BI561792.1 | - | 3 | 324 | |||
| BI826726.1 | - | 3 | 284 | |||

TMCO5A
Genomic Map