siRNA Details
| Gene Symbol: ZNF362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152493.2 | NP_689706.2 | 9 | 2218 | ||
| XM_005270507.2 | XP_005270564.1 | 8 | 2358 | |||
| XM_005270508.4 | XP_005270565.1 | 9 | 3015 | |||
| XM_017000415.1 | XP_016855904.1 | 8 | 2930 | |||
| GenBank mRNA | AF318356.1 | - | 8 | 2068 | ||
| AK098342.1 | - | 1 | 458 | |||
| BC071632.1 | - | 2 | 1030 | |||
| BC103704.1 | - | 1 | 651 | |||

ZNF362
Genomic Map