siRNA Details
| Gene Symbol: CNIH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322302.1 | NP_001309231.1 | 3 | 390 | ||
| NM_001322303.1 | NP_001309232.1 | 2 | 324 | |||
| NM_001322304.1 | NP_001309233.1 | 3 | 468 | |||
| NM_001322305.1 | NP_001309234.1 | 2 | 810 | |||
| NM_152495.1 | NP_689708.1 | 2 | 810 | |||
| NR_136287.1 | 2 | 810 | ||||
| NR_136288.1 | 3 | 320 | ||||
| NR_136289.1 | 4 | 456 | ||||
| NR_136290.1 | 3 | 390 | ||||
| NR_136291.1 | 4 | 526 | ||||
| NR_136292.1 | 3 | 460 | ||||
| NR_136293.1 | 6 | 696 | ||||
| NR_136294.1 | 5 | 630 | ||||
| NR_136295.1 | 4 | 501 | ||||
| NR_136296.1 | 4 | 526 | ||||
| NR_136297.1 | 3 | 390 | ||||
| GenBank mRNA | AF070524.1 | - | 1 | 16 | ||
| AK096312.1 | - | 2 | 810 | |||
| BC022780.1 | - | 2 | 298 | 3894832 | ||

CNIH3
Genomic Map