siRNA Details
| Gene Symbol: LINC00479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027272.1 | 5 | 517 | |||
| GenBank mRNA | AK057397.1 | - | 5 | 517 | ||
| BC128198.1 | - | 5 | 538 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00479| Gene Symbol: LINC00479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027272.1 | 5 | 517 | |||
| GenBank mRNA | AK057397.1 | - | 5 | 517 | ||
| BC128198.1 | - | 5 | 538 | |||